Potassium lactate solution 60%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassium 2-hydroxypropanoate
, Lactic acid, potassium salt
P1806-1KG
P1806-50GM
P1806-200GM
700015-194EA
205.35
USD
700015-194
700015-196
700015-192
Potassium lactate solution 60%
Potassium lactate
Potassium Lactate, 60 Percent Solution is used in personal care due to its high moisturizing properties as well as for its buffering capacity.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₃H₅O₃K MW: 128.168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036410 CAS Number: 996-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...