Poly(diallyldimethylammonium chloride) [pDADMAC] 35% in aqueous solution, very low molecular weight

Synonyms: Polyquaternium-6PolyDADMAC

P2818-250ML
700012-820EA 204 USD
700012-820
Poly(diallyldimethylammonium chloride) [pDADMAC] 35% in aqueous solution, very low molecular weight
Poly(diallyldimethylammonium chloride) [pDADMAC]
Formula: (C₈H₁₆ClN)n
Density: 1.09
Storage Temperature: Ambient
MDL Number: MFCD00192409
CAS Number: 26062-79-3

Order Now

Specification Test Results

Viscosity @ 25°C 100-200 cPs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR