SPCMP1867
P1867-2.5LTG P1867-500MLG
700012-654EA 132.15 USD
700012-654 700012-652
SPCMP1867
Polyethylene Glycol 400 Monolaurate, commonly referred to as PEG 400 MONOLAURATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available
GHS hazard : Exclamation mark