SPCMP2129
P2129-450LBB P2129-18KG P2129-500GM P2129-2.5KG
700015-340EA 2873.61 USD
700015-340 700015-338 700015-336 700015-342
SPCMP2129
Polyethylene Glycol 25 Castor Oil, Hydrogenated is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available