SPCMP2830
P2830-500GM
700012-688EA 61.02 USD
700012-688
SPCMP2830
POLYETHYLENE GLYCOL 200 MONOOLEATE, commonly referred to as PEG 200 MONOOLEATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.