Poly(methyl acrylate) solution 40% in toluene

Synonyms: Acrylic Acid Methyl Ester

P2892-50GM P2892-10GM P2892-100GM
700012-382EA 1046.6 USD
700012-382 700012-386 700012-384
Poly(methyl acrylate) solution 40% in toluene
Poly(methyl acrylate)

Poly(Methyl Acrylate), 40 Percent Solution in Toluene, also known as Polymethylacrylate or Polymethyl acrylate, is a hydrophobic synthetic acrylate polymer used in textiles and leather finishing. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability


UN1294

Formula: (C₄H₆O₂)n
Density: 1,22
Storage Temperature: Ambient
MDL Number: MFCD00084441
CAS Number: 9003-21-8
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Assay Approx. 40%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR