Pimenta leaf oil

Supplier: SPECTRUM CHEMICAL MFG CORP

P1199-25ML P1199-125ML P1199-500MLG
700015-070EA 458 USD
700015-070 700015-066 700015-068
Pimenta leaf oil
Pimenta leaf oil

Pimenta Leaf Oil is a flavor and fragrance ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 1.044
Storage Temperature: Ambient
CAS Number: 8016-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR