Phosphorus, red, amorphous fine powder (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP

P1110-25GM P1110-100GM P1110-6X500GM P1110-500GM
10841-956CS 502 USD
10841-956 10841-954 10841-952 10841-950
Phosphorus, red, amorphous fine powder (DEA List I Chemical)
Phosphorus

Phosphorus, Red (DEA List I Chemical), Amorphous, Fine Powder, a chemical element, is widely used in fertilizers as an essential plant nutrient. It is also used to make organophosphorus compounds. The United States Drug Enforcement Agency (DEA) maintains a list of illicit drugs and the chemical materials used to synthetically produce them. List I chemicals are those that are used in the manufacture of the controlled substances and are important to the manufacture of the substances. Regulatory forms are required to purchase this material


GHS hazard : Flame;Exclamation mark
UN1338

Formula: P
MW: 30.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
UN: 1338
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR