Phenyltoloxamine citrate

P1774-25KGBL P1774-25GM
700007-474EA 140.84 USD
700007-474 700007-476
Phenyltoloxamine citrate
Phenyltoloxamine citrate

Phenyltoloxamine Citrate, an antihistamine with analgesic and sedative effects, is used as an enhancing agent for some antitussives and sometimes as cough suppressant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₂₁NO·C₆H₈O₇
Storage Temperature: Ambient
MDL Number: MFCD00038875
CAS Number: 1176-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR