(R)-(−)-Phenylephrine hydrochloride ≥98.0% USP

Synonyms: (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol hydrochloride(R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol hydrochlorideL(-)-Phenylephrine hydrochloride(R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol hydrochlorideR)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride(R)-Phenylephrine hydrochloride

PH147-5KGBL PH147-5GM PH147-25GM PH147-1KGBL PH147-100GM
95034-930EA 735.34 USD
95034-930 95034-936 95034-934 700015-476 95034-932
(R)-(−)-Phenylephrine hydrochloride ≥98.0% USP
(R)-(−)-Phenylephrine hydrochloride

Phenylephrine Hydrochloride, USP is used for the temporary relief of stuffy nose, sinus, and ear symptoms caused by the common cold, flu, and allergies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₉H₁₃NO₂·HCl
Storage Temperature: Ambient
MDL Number: MFCD00012605
CAS Number: 61-76-7

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation @25 C -43° - -47°
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.2 %
Sulfate max. 0.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR