SPCMPH148

PH148-25GM PH148-100GM PH148-500GM PH148-1KG
95034-928EA 515.61 USD
95034-928 95034-926 95034-924 95034-922
SPCMPH148
Phenylbutazone, USP is a nonsteroidal anti-inflammatory drug for treating fever and pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR