Petitgrain oil FCC Paraguay Type
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Petitgrain Oil, Paraguay Type, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark
Storage Temperature:
Ambient |
MDL Number:
MFCD00217648 CAS Number: 8014-17-3 |
Specification Test Results
Assay | 45.0 - 60.0 % |
Angular Rotation | -4 to +1 |
Refractive Index @ 20°C | 1.455- 1.462 |
Solubility in Alcohol | To Pass Test |
Specific Gravity @ 25°C | 0.878- 0.889 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...