Pentafluoropropionic acid ≥98%

Synonyms: Perfluoropropionic acidPerfluoropropanoic acid2,2,3,3,3-Pentafluoropropionic acidPentafluoropropanoic acid

P1201-25GM P1201-100GM
10841-834EA 125.8 USD
10841-834 10841-836
Pentafluoropropionic acid ≥98%
Pentafluoropropionic acid

Caution: Storage temperature - HYGROSCOPIC: Keep tightly closed in a cool, dry place

Formula: C₃HF₅O₂
MW: 164.03 g/mol
Density: 1.561
Storage Temperature: Ambient
MDL Number: MFCD00004170
CAS Number: 422-64-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Purity (GC) 98 %
Purity (Neutralization Titration) 98 %
Specific Gravity (20/20) 1.5760 - 1.5800

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR