Palmitoleic acid ≥99%

Synonyms: (Z)-Hexadec-9-enoic acid

P3601-5GM P3601-1GM P3601-100MG
10860-348EA 1402.3 USD
10860-348 10860-344 10860-346
Palmitoleic acid ≥99%
Palmitoleic acid

Palmitoleic Acid, also known as cis-Palmitoleic acid or 9-cis-Hexadecenoic acid, is an omega-7 monounsaturated fatty acid that can increase insulin sensitivity by suppressing inflammation.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
Density: 0.895
Storage Temperature: Refrigerator
MDL Number: MFCD00004437
CAS Number: 373-49-9

Order Now

Specification Test Results

Assay Approx. 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR