Palm oil
Supplier: SPECTRUM CHEMICAL MFG CORP
PA105-2.5KG
PA105-200LTB
PA105-500GM
700000-120EA
372
USD
700000-120
700000-122
700000-124
Palm oil
Palm oil
Palm Oil is a vegetable oil that is edible and comes from the mesocarp of the oil palms. It can also be used to make biodiesel.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00147822 CAS Number: 8002-75-3 |
Specification Test Results
Iodine Value | 46 - 55 |
Free Fatty Acids (As Oleic Acid) | max. 0.1% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...