Oxymetazoline hydrochloride ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochlorideAfrin hydrochlorideSOxymetazoline hydrochloride

O1359-5GM O1359-25GM O1359-500GMB O1359-100GM
95031-352EA 1049 USD
95031-352 95031-354 95031-350 95031-356
Oxymetazoline hydrochloride ≥98.5% USP
Oxymetazoline hydrochloride

Oxymetazoline Hydrochloride, USP is used for temporary relief of congestion in the nose caused by various conditions including the common cold, sinusitis, hay fever, and allergies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₆H₂₄N₂O·HCl
MW: 296.839 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058147
CAS Number: 2315-02-8
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
pH 4.0 - 6.5
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR