Orange oil FCC, cold-pressed California Type

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Citrus sinensisCitrus aurantium

O1012-20LTBL O1012-200LTBL O1012-500MLG O1012-125ML
75811-758EA 5061 USD
75811-758 700010-894 75811-760 95030-766
Orange oil FCC, cold-pressed California Type
Orange oil

Orange Oil, California Type, Coldpressed, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark
UN1197

Storage Temperature: Ambient
MDL Number: MFCD00147798
CAS Number: 8028-48-6
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Assay [Aldehydes As Decyl Aldehyde (C₁₀H₂₀O] 1.2 - 2.5 %
Angular Rotation +94 to +99
Refractive Index @ 20°C 1.472 - 1.474
Specific Gravity @ 25°C 0.842 - 0.846
Ultraviolet Absorbance 0.130 min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR