Opopanax oil

O1297-45KGBL
700010-752EA 36658.46 USD
700010-752
Opopanax oil
Opopanax oil

Opopanax Oil is a fragrance ingredient with a balsamic type odor.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.900
Storage Temperature: Ambient
MDL Number: MFCD00678435
CAS Number: 8021-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR