2-Ethylhexyl-4-methoxycinnamate
Synonyms:
Octinoxate
, 4-Methoxycinnamic Acid Octyl Ester
, Octyl 4-Methoxycinnamate
, 4-Methoxycinnamic Acid 2-Ethylhexyl Ester
, 2-Ethylhexyl 3-(4-methoxyphenyl)acrylate
Octyl Methoxycinnamate, also known as Ethylhexyl methoxycinnamate, is used as an ingredient in lip balms and sunscreens to absorb UV-B rays from the sun, protecting the skin from damage and reduces the appearance of scars.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₈H₂₆O₃ MW: 290.40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00072582 CAS Number: 5466-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...