2-(1-Naphthylmethyl)-2-imidazoline hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: AlbalonNaphazoline Hydrochloride

NA106-100GM NA106-5KGBL NA106-25GM NA106-5GM NA106-1KGBL
700008-670EA 34918 USD
700008-670 95034-696 95034-694 95034-692 95034-690
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride ≥98.0% USP
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride

Naphazoline Hydrochloride, USP is a decongestant used to relieve typical allergy symptoms. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Skull and crossbones
UN2811

Formula: C₁₄H₁₅ClN₂
MW: 246.738 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012554
CAS Number: 550-99-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 -102.0 %
pH of a 1 % Solution @ 25°C 5.0 - 6.6
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.2 %
Ordinary Impurities To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR