N,N'-Ethylenebis(stearamide)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N,N'-1,2-EthanediylbisoctadecanamideEthylene DistearamideHoechst wax C

E1179-5KG E1179-250GM E1179-1KG
700007-756EA 372.42 USD
700007-756 700007-754 700007-752
N,N'-Ethylenebis(stearamide)
N,N'-Ethylenebis(stearamide)
Formula: C₃₈H₇₆N₂O₂
MW: 593,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059224
CAS Number: 110-30-5

Order Now

Specification Test Results

Appearance White to Off-Whi
Elemental Analysis Carbon 75.0 - 78.0%
Nitrogen 4.5 - 4.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR