N,N-Diethylformamide ≥99.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: FormdiethylamideFormic acid diethylamide

D2358-25ML D2358-500ML D2358-100ML
10841-832EA 351.97 USD
10841-832 10841-828 10841-830
N,N-Diethylformamide ≥99.0%
N,N-Diethylformamide

GHS hazard : Flame;Exclamation mark
UN1993

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Purity (GC) 99.0 %
Specific Gravity (20/20) 0.9060 - 0.9080
Refractive Index (N20/D) 1.4330 - 1.4360

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR