N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥99%

Synonyms: N-(2-Hydroxyethyl)ethylenediaminetriacetic acid2,2-((2-((Carboxymethyl)(2-hydroxyethyl)amino)ethyl)azanediyl)diacetic acid

H1111-500GM H1111-2.5KG
700007-460EA 827.1 USD
700007-460 700007-462
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥99%
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid

N-(2-Hydroxyethyl)ethylenediaminetriacetic Acid is a metal and trace metal chelating agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₈N₂O₇
MW: 278.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004294
CAS Number: 150-39-0

Order Now

Specification Test Results

Assay Approx. 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR