Musk ketone ≥95%

M1345-500GM M1345-25GM M1345-125GM
700004-208EA 932.25 USD
700004-208 700004-206 700004-204
Musk ketone ≥95%
Musk ketone

Musk Ketone, also known as Synthetic musk or white musk, is a synthetic aromachemical used to emulate the scent of deer musk in perfumery, cosmetics, and detergents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₁₈N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00024271
CAS Number: 81-14-1

Order Now

Specification Test Results

Assay 95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR