Morphine sulfate ≥98.0% (CII) USP

Supplier: SPECTRUM CHEMICAL MFG CORP

M1167-500GMBL M1167-25GM M1167-1GM M1167-5GM M1167-100GM
75811-586EA 142 USD
75811-586 75811-588 75811-584 75811-590 75811-592
Morphine sulfate ≥98.0% (CII) USP
Morphine sulfate

Morphine Sulfate (CII), USP is an opioid analgesic. It is a DEA CONTROLLED SUBSTANCE - SCHEDULE II and does require DEA registration. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: (C₁₇H₁₉NO₃)₂·H₂SO₄·5H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150902
CAS Number: 6211-15-0

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0 - 102.0 %
Specific Rotation [A]D -107° to -109.5
Water Determination 10.4 - 13.4 %
Acidity To Pass Test
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR