SPCMM1198
M1198-500MLG
95034-628EA 100.79 USD
95034-628
SPCMM1198
MINERAL OIL, WHITE, HEAVY, FCC is the by product of the distillation of petroleum, it is most often translucent, odorless and produced in very large quantities. The use of food grade mineral oil is self-limiting because of its laxative effect. However, it is used as a popular preservative for wooden cutting boards, salad bowls and utensils because of its properties that prevent water absorption, combined with its lack of flavor and odor. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers