Miconazole nitrate ≥98.0% USP

Synonyms: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate(±)-Miconazole nitrate salt

MI150-500GM MI150-1KGBL MI150-100GM MI150-50KGBL MI150-25GM MI150-5GM MI150-1GM MI150-5KGBL
95034-578EA 2154.52 USD
95034-578 75811-734 95034-576 95034-574 75811-732 95034-582 95034-580 700008-668
Miconazole nitrate ≥98.0% USP
Miconazole nitrate

Miconazole Nitrate, USP is a topical imidazole antifungal agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
MW: 479.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058161
CAS Number: 22832-87-7

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.2 %
Related Compounds To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR