Methyleugenyl PEG-8 dimethicone
Supplier: SPECTRUM CHEMICAL MFG CORP
M1702-1KG
M1702-100GM
M1702-25GM
700014-834EA
110
USD
700014-834
700014-832
700014-830
Methyleugenyl PEG-8 dimethicone
Methyleugenyl PEG-8 dimethicone
Methyleugenyl PEG-8 Dimethicone is used in hair and skin conditioning products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
CAS Number:
200443-93-2 |
Specification Test Results
Appearance | Clear Liquid |
Refractive Index | Approx. 1.505 |
Viscosity | 1000 - 2000 cSt |
Specific Gravity | Approx. 1.09 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...