Diiodomethane ≥98% stabilized
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Methylene iodide
Stabilization:
Stabilised with copper
M1258-100GM
M1258-25GM
M1258-500GM
10841-868EA
168
USD
10841-868
10841-870
10841-872
Diiodomethane ≥98% stabilized
Diiodomethane
Methylene Iodide, Stabilized with Copper, also known as Diiodomethane, can be used to determine the density of mineral and solid samples. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
GHS hazard : Exclamation mark
UN2810
Formula:
CH₂I₂ MW: 267.84 g/mol Density: 3.325 Storage Temperature: Ambient |
MDL Number:
MFCD00001079 CAS Number: 75-11-6 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...