Methyl-β-cyclodextrin (mixture of several Methylated)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methylated-β-Cyclodextrin

M2799-250GMB M2799-10GM M2799-5GM
700011-328EA 1187 USD
700011-328 700011-326 700011-330
Methyl-β-cyclodextrin (mixture of several Methylated)
Methyl-beta-cyclodextrin (mixture of several Methylated)

Methyl-beta-cyclodextrin, also known as Cyclodextrins, can form complexes with hydrophobic compounds with use in the food and pharmaceutical industries as a solubilizer, stabilizer, emulsifier and odor reducing agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

MW: 1303.3000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074980
CAS Number: 128446-36-6

Order Now

Specification Test Results

Specific Rotation [A]D +160 +/- 5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR