Methyl-4-aminobenzoate

Synonyms: Methyl 4-aminobenzoate

M1076-25GM M1076-100GM
10860-582EA 314.33 USD
10860-582 10860-584
Methyl-4-aminobenzoate
Methyl 4-aminobenzoate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007891
CAS Number: 619-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR