Hexamethylenetetramine ≥99.0% USP

Synonyms: HexamineHexamethylenetetramineUrotropine

M1447-12KG M1447-50KGBL M1447-100KGBL
75811-664EA 7265.88 USD
75811-664 700008-968 700008-966
Hexamethylenetetramine ≥99.0% USP
Methenamine

Methenamine, USP is an antibiotic is used to prevent or control returning urinary tract infections. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark;Health hazard
UN1328

Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
UN: 1328
ADR: 4.1,III
https://us.vwr.com/stibo/web/std.lang.all/84/97/2298497.jpg

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5%
Loss On Drying max. 2.0%
Residue On Ignition max. 0.1%
Chloride (Cl) max. 0.014%
Sulfate To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR