5-Aminosalicylic acid ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5-Amino-2-hydroxybenzoic acidMesalamineMesalazinem-Aminosalicylic acid

M1540-500GMG M1540-2.5KGP M1540-25GMGL M1540-100GMG
95031-570EA 878 USD
95031-570 95031-568 95031-564 95031-566
5-Aminosalicylic acid ≥98.5% USP
5-Aminosalicylic acid

Mesalamine, USP is used primarily as an anti-inflammatory drug in the treatment of ulcerative colitis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007877
CAS Number: 89-57-6

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Clarity of Solution To Pass Test
pH (1 in 40 Suspension) 3.5 - 4.5
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR