Meldola's Blue

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 8-Dimethylamino-2,3-benzophenoxazine hemi (zinc chloride) salt9-(Dimethylamino)benzo[a]phenoxazin-7-ium chloride zinc chloride

M3807-5GM
700014-862EA 353 USD
700014-862
Meldola's Blue
Meldola's Blue
Formula: C₁₈H₁₅ClN₂O·Cl₂Zn
Storage Temperature: Ambient
MDL Number: MFCD00012113
CAS Number: 7057-57-0

Order Now

Specification Test Results

Absorption Max 570 NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR