Manganese gluconate ≥98.0%, dried powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Manganesegluconate

M1112-500GM M1112-2.5CS4 M1112-500CS6 M1112-2.5KG-CS M1112-500GM-CS M1112-50KGBL M1112-12KG M1112-2.5KG
95034-304CS 1805 USD
95034-304 76014-610 89078-874 95034-310 700006-254 76014-608 95034-308 95034-306
Manganese gluconate ≥98.0%, dried powder USP
Manganese gluconate

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂MnO₁₄
MW: 445.2327 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00867656
CAS Number: 6485-39-8

Order Now

Specification Test Results

Assay (C₁₂H₂₂Mno₁₄;Anhyd.Basis) 98.0 - 102.0 %
Water 3.0 - 9.0 %
Chloride (Cl) max. 0.05 %
Sulfate max. 0.2 %
Lead (Pb) max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR