Magnesium(II) gluconate hydrate ≥98.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
D-Gluconic acid magnesium salt hydrate
, D-Gluconic acid hemimagnesium salt hydrate
, Magnesium D-gluconate hydrate
, Gluconic acid magnesium(II) salt hydrate
Magnesium Gluconate, USP is an essential mineral and used in treating low magnesium levels or maintaining the proper amount of magnesium in the body. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₁₂O₇·xH₂O MW: 414.60 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150971 CAS Number: 3632-91-5 |
Specification Test Results
Assay (C₁₂H₂₂MGO₁₄; Anhydrous Basis | 98.0 - 102.0 % |
pH of a 5% Solution | 6.0 - 7.8 |
Water | 3.0 - 12.0 % |
Chloride (Cl) | max. 0.05 % |
Sulfate | max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...