Magnesium carbonate hydrate, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Magnesium carbonate hydrate

MA108-2.5KG MA108-12KG MA108-2.5CS4 MA108-500CS6 MA108-20KGBL
700000-400EA 868.76 USD
700000-400 95034-174 95034-172 95034-170 75811-690
Magnesium carbonate hydrate, powder USP
Magnesium carbonate hydrate

Magnesium Carbonate, Light Powder, USP is used as a directly compressible tablet diluent in concentrations up to 45%. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgCO₃·xH₂O
MW: 84.31 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151022
CAS Number: 23389-33-5

Order Now

Specification Test Results

Assay (MgO) 40.0 - 43.5 %
Microbial Limits: To Pass Test
Soluble Salts max. 1.0 %
Acid-Insoluble Substances max. 0.05 %
Arsenic (As) max. 4 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR