(-)-Menthyl acetate ≥97.5%

Synonyms: (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate(R)-(-)-Menthyl acetate(-)-Acetoxy-p-menthane(5R)-2-Isopropyl-5-methylcyclohexyl acetate

M1660-25ML M1660-100ML
700004-548EA 90.62 USD
700004-548 700004-546
(-)-Menthyl acetate ≥97.5%
(-)-Menthyl acetate

L-Menthyl Acetate, also known as (1R)-()-Menthyl acetate, is a natural monoterpene and the acetate ester of menthol; it has a peppermint scent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Density: 0.92
Storage Temperature: Ambient
MDL Number: MFCD00001482
CAS Number: 2623-23-6

Order Now

Specification Test Results

Assay 97.5% min.
Optical Rotation -79° to -83°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR