Phenol liquified ≥89% (w/w)

P1592-500ML P1592-4LTPV P1592-2.5LTPV
10841-812EA 843 USD
10841-812 10841-810 10841-814
Phenol liquified ≥89% (w/w)
Phenol liquified

Storage temperature - LIGHT AND AIR SENSITIVE: Keep tightly closed in light-resistant containers


GHS hazard : Skull and crossbones;Corrosion;Health hazard
UN2821

Formula: C₆H₅OH
MW: 94.11 g/mol
Density: 1.071
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 2821
ADR: 6.1,II

Order Now

Specification Test Results

Assay (C₆H₆O); By Wt. 89.0 % min.
Clarity of Solution and Reaction To Pass Test
Stabilizer As Reported
Distilling Range max. 182.5°C
Nonvolatile Residue max. 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR