Linseed oil (raw)

Supplier: SPECTRUM CHEMICAL MFG CORP

LI113-4LTGL LI113-20LT LI113-500MLG
700000-008EA 989.63 USD
700000-008 700000-010 700000-012
Linseed oil (raw)
Linseed oil

Linseed Oil, Raw, also known as flaxseed oil, is used as an impregnator and varnish in wood finishing because of its polymer-forming properties. It can also be used as a nutritional supplement or as a pigment binder in oil paints.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.930
Storage Temperature: Ambient
MDL Number: MFCD00131507
CAS Number: 68553-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR