Linseed oil, boiled
Supplier: SPECTRUM CHEMICAL MFG CORP
LI111-4LTGL
LI111-20LT
LI111-200LTB
LI111-500MLG
700000-004EA
436
USD
700000-004
700000-006
700000-000
700000-002
Linseed oil, boiled
Linseed oil
Linseed Oil, Boiled, also known as flaxseed oil, is used as an impregnator and varnish in wood finishing because of its polymer-forming properties. It can also be used as a nutritional supplement or as a pigment binder in oil paints.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
0.930 Storage Temperature: Ambient |
MDL Number:
MFCD00131507 CAS Number: 68553-15-1 |
Specification Test Results
Appearance | Yellow to Brown |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...