Lidocaine hydrochloride monohydrate ≥97.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

LI103-25GM LI103-10KGBL LI103-50KGBL LI103-25KGBL LI103-5KG LI103-500GM LI103-100GM LI103-5GM
95033-978EA 79 USD
95033-978 75811-536 75811-538 75811-540 95033-972 89078-838 95033-974 95033-976
Lidocaine hydrochloride monohydrate ≥97.5% USP
Lidocaine hydrochloride monohydrate

Lidocaine Hydrochloride, Monohydrate, USP is used as a topical local anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₁₄H₂₃ClN₂O·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150329
CAS Number: 6108-05-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.5 - 102.5%
Water Determination 5.0 - 7.0%
Residue After Ignition max. 0.1%
Sulfate max. 0.1 %
Heavy Metals (As Pb) max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR