Lidocaine ≥97.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

LI102-5KG LI102-100GM LI102-500GM LI102-25GM LI102-25KGBL
75811-534EA 2643 USD
75811-534 95033-980 95033-982 89078-840 95033-984
Lidocaine ≥97.5% USP
Lidocaine

Lidocaine, USP, also known as xylocaine or lignocaine is used as a topical local anesthetic and medication to numb tissue in a specific area. It also has uses as a treatment for ventricular tachycardia and to perform nerve blocks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₁₄H₂₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00026733
CAS Number: 137-58-6

Order Now

Specification Test Results

Assay (C₁₄H₂₂N₂O) 97.5 - 102.5 %
Melting Range 66 - 69 °C
Chloride (Cl) max. 0.0035 %
Residue On Ignition max. 0.1 %
Sulfate max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR