3-(3,4-Dihydroxyphenyl)-L-alanine ≥98.0%, Powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: DoparL-dopa3-Hydroxy-L-tyrosineLevodopaL-β-3,4-Dihydroxyphenyl-Alanine

D1066-25GM D1066-500GM D1066-100GM D1066-5GM
95033-914EA 1156 USD
95033-914 95033-916 95033-918 95033-920
3-(3,4-Dihydroxyphenyl)-L-alanine ≥98.0%, Powder USP
3-(3,4-Dihydroxyphenyl)-L-alanine

Levodopa, Powder, USP is used to treat the stiffness, tremors, spasms, and poor muscle control of Parkinson's disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0%
Specific Rotation [A]D -160° to -167°
Loss On Drying max. 0.5%
Residue On Ignition max. 0.1%
Heavy Metals max. 0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR