Lemon oil FCC, cold-pressed California Type
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Lemon Oil, California Type, Cold-pressed, FCC is used in aromatherapy and as a flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame;Health hazard;Exclamation mark
UN1197
Storage Temperature:
Ambient |
MDL Number:
MFCD00147430 CAS Number: 8008-56-8 UN: 1197 ADR: 3,III |
Specification Test Results
Assay (Aldehydes As Citral) | 2.2 - 3.8 % |
Angular Rotation | +57 to +65.6 |
Refractive Index @ 20°C | 1.473 - 1.476 |
Solubility in Alcohol | To Pass Test |
Specific Gravity @ 25°C | 0.849 - 0.855 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...