(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(S)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-hydroxypropanoate
, L-Menthyl L-Lactate
, (S)-2-Hydroxypropionic acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester
M1680-5GM
M1680-25GM
700014-822EA
87
USD
700014-822
700014-820
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
Formula:
C₁₃H₂₄O₃ MW: 228.33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27977194 CAS Number: 61597-98-6 |
Specification Test Results
Appearance | White Crystallin |
Melting Point | 40°C min. |
Assay (GC) | 97% min. |
Optical Rotation | -79° to -83° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...