Jasmine oil

Supplier: SPECTRUM CHEMICAL MFG CORP

J1010-500MLG J1010-100ML
700010-668EA 434 USD
700010-668 700010-666
Jasmine oil
Jasmine oil

Jasmine Oil is a proprietary blend of natural and synthetic jasmine oils. While the pure extract is used in cosmetic and personal care product formulations as a fragrance ingredient for its masking, moisturizing, skin conditioning, and soothing properties, the blended product is not.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.947
Storage Temperature: Ambient
MDL Number: MFCD00147303
CAS Number: 8022-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR