Ichthammol USP

Supplier: SPECTRUM CHEMICAL MFG CORP

IC110-12KG IC110-125GM IC110-500GM IC110-50KGBL IC110-2.5KG
700014-592EA 9188 USD
700014-592 700014-590 95033-672 95033-674 95033-676
Ichthammol USP
Ichthammol

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 8029-68-3

Order Now

Specification Test Results

Ammonia Content (NH ) 2.5 % min.
Ammonia Content (NH3) 2.5 % min.
Sulfur Content Total (S) 10.0 % min.
Sulfur Content Total (S) 10.0 % min.
Loss On Drying max. 50.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR