SPCMHY119
HY119-125GM HY119-12KG HY119-500GM HY119-2.5KG
700013-024EA 3393.68 USD
700013-024 CA95033-584 95033-586 95033-588
SPCMHY119
Hydroxypropyl Cellulose, 150-400 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications.
Certifications: All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.