Hydromorphone hydrochloride ≥98.0% (CII) USP

H1107-10GM H1107-5GM H1107-1GM H1107-25GM H1107-100GMBL
75811-328EA 8200.79 USD
75811-328 75811-334 75811-336 75811-330 75811-332
Hydromorphone hydrochloride ≥98.0% (CII) USP
Hydromorphone hydrochloride

Hydromorphone Hydrochloride (CII), USP Hydromorphone Hydrochloride (CII), USP is the water soluble salt of hydromorphone, an opioid and analgesic drug. It is legally considered a narcotic and commonly used in hospitals to act on the central nervous system and gastrointestinal tract. It is a DEA CONTROLLED SUBSTANCE - SCHEDULE II drug and does require DEA registration by the customer and completion of DEA Form 222. Certification of Available Procurement Quota required for certain customers. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₇H₁₉NO₃·HCl
Storage Temperature: Ambient
MDL Number: MFCD03427563
CAS Number: 71-68-1

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 101.0 %
Specific Rotation [A]D -136 to -139
Acidity To Pass Test
Loss On Drying max. 1.5 %
Residue On Ignition max. 0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR